Pycnometer
5 ml
Katalog # HECH42570008
Leverantör: GLASWARENFABRIK KARL HECHT
Gå till:
Produktdetaljer och dokument
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in stopper
- Good chemical resistance
- Good temperature resistance
Certifications: DIN ISO 3507
Specifikationer
- Volym (nominell):5 ml