Micro pipettes, disposable, with ring mark
50 µl, Grön, En markering
Katalog # HECH40555021
Leverantör: GLASWARENFABRIK KARL HECHT
Micro pipettes, disposable, with ring mark
Katalog # HECH40555021
Leverantör: GLASWARENFABRIK KARL HECHT
Gå till:
Product Details & Documents
Disposable micro pipettes with ring mark.
- Calibrated to contain 'In'
- Colour-coded
Certifications: DIN ISO 7550
Specifikationer
- Färgkod:Grön
- Typ:En markering
- Kapacitet:50 µl
- Seq:04