Intravenös kateter, Surflash™
Intravenös kateter, G 22, Blå, PU, 25 mm
Katalog # TERUSR-FF2225
Leverantör: TERUMO
Gå till:
Produktdetaljer och dokument
Surflash is made of PU, which is firm at room temperature but soft at body temperature, making the catheter more comfortable.
- Only exists in the straight version
- Enables quick and easy confirmation of the placement in the vein by an additional flashback
- Higher certainty of proper placement
Helpful with patients whose veins are hard to detect and in which venipuncture is more difficult, such as small children, people undergoing chemotherapy.
Dokument
Specifikationer
- Mätare:G 22
- Färg:Blå
- Beskrivning:Intravenös kateter
- Längd:25 mm
- Material:PU