Intravenös kateter, Surflash™
Intravenös kateter, G 20, Rosa, PU, 32 mm
Katalog # TERUSR-FF2032
Leverantör: TERUMO
Gå till:
Produktdetaljer och dokument
Surflash is made of PU, which is firm at room temperature but soft at body temperature, making the catheter more comfortable.
- Only exists in the straight version
- Enables quick and easy confirmation of the placement in the vein by an additional flashback
- Higher certainty of proper placement
Helpful with patients whose veins are hard to detect and in which venipuncture is more difficult, such as small children, people undergoing chemotherapy.
Specifikationer
- Mätare:G 20
- Färg:Rosa
- Beskrivning:Intravenös kateter
- Längd:32 mm
- Material:PU
- Seq:005