Orders LinkContactUsLinkComponent
Intravenös kateter, Surflash™
Intravenös kateter, Surflash™
Intravenös kateter, G 16, Grå, PU, 51 mm
Katalog # TERUSR-FF1651
Leverantör:   TERUMO
Intravenös kateter, Surflash™
Katalog # TERUSR-FF1651
Leverantör:  TERUMO
Intravenös kateter, Surflash™
Intravenös kateter, G 16, Grå, PU, 51 mm
Price
1 275,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 50 Items
Product Details & Documents
Surflash is made of PU, which is firm at room temperature but soft at body temperature, making the catheter more comfortable.
  • Only exists in the straight version
  • Enables quick and easy confirmation of the placement in the vein by an additional flashback
  • Higher certainty of proper placement
Helpful with patients whose veins are hard to detect and in which venipuncture is more difficult, such as small children, people undergoing chemotherapy.
Specifikationer
  • Mätare:
    G 16
  • Färg:
    Grå
  • Beskrivning:
    Intravenös kateter
  • Längd:
    51 mm
  • Material:
    PU
  • Seq:
    002