Dissecting scissors
Curved, sharp/sharp, 115 mm
Katalog # HAMML-HSB301-11
Leverantör: HAMMACHER KARL
Gå till:
Product Details & Documents
These dissecting scissors made of stainless steel feature tungsten carbide inserts.
Specifikationer
- Typ:Curved, sharp/sharp
- Längd:115 mm
- Seq:02