Buffert, bruksfärdig
Technical buffer TPL-4/25, 4,00, Plastic bottle, 25
Catalog # WTWA108811
Supplier: WTW
Jump to:
Product Details & Documents
Every bottle has its own lot reference with the expiration date.
- Shelf life up to 3 years (unopened)
- Reference temperature at 25 °C
- Economic consumption
- Easiest handling by use of the dosing vessel
- One litre container with a separate compartment for the sample
Accuracy is ±0,03 pH
Certifications: Traceable to NIST.
Specifications
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:25
- Beskrivning:Technical buffer TPL-4/25
- pH-värde:4,00



