Plugs for test tubes
12 mm, Grön
Katalog # 212-0527
Leverantör: BOETTGER PAUL
Gå till:
Produktdetaljer och dokument
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
- Not autoclavable
- Easy handling with a high, ribbed grip
- Easy to open by tilting 'one-hand operation'
Specifikationer
- Färg:Grön
- Seq:05
- Storlek:12 mm