Orders LinkContactUsLinkComponent
CellStor Pot™ specimen container
CellStor Pot™ specimen container
90 ml, 10% formal saline
Katalog # 720-2325
Leverantör:   CELLPATH
CellStor Pot™ specimen container
Katalog # 720-2325
Leverantör:  CELLPATH
CellStor Pot™ specimen container
CellStor Pot™ specimen container
90 ml, 10% formal saline
Price
325,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 25 Items
Produktdetaljer och dokument
Prefilled specimen containers.
  • Leak-proof lid for safe transportation and specimen storage
  • With traceable badge number, safety data sheet in the packaging
Specifikationer
  • Kapacitet:
    90 ml
  • Beskrivning:
    10% formal saline
  • Seq:
    06