Accessories for data loggers, Dickson®
DicksonWare™ software and USB cable, for data loggers
Katalog # 620-1515
Leverantör: DICKSON
Gå till:
Product Details & Documents
DicksonWare™ software (incl. USB or serial cable) is needed for setting up Dickson loggers. They enable viewing and downloading data, and resetting the logger to the factory settings.
Specifikationer
- Beskrivning:DicksonWare™ software and USB cable, for data loggers
- Cat. No.:620-1515
- Gränssnitt:USB