Solid phase microextraction fibers
StableFlex fiber kit: PDMS/DVB, DVB/carboxen/PDMS, carboxen/PDMS and polyacrylate, for manual use
Katalog # VARISU57550U
Leverantör: VARIAN
Gå till:
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Specifikationer
- Beskrivning:StableFlex fiber kit: PDMS/DVB, DVB/carboxen/PDMS, carboxen/PDMS and polyacrylate, for manual use
- Seq:31
Accessories
Produktinformation
- Katalog #TillgänglighetPrice
- Merlin microseal, SPME replacement seal, 23 gaugeMerlin microseal, SPME replacement seal, 23 gaugeVARI392609902Log in för att kolla tillgänglighet och ert pris1 * 1 Items - 10 511,32 kr
Produktinformation
- Katalog #TillgänglighetPrice
- SPME fiber holder for CTC autosamplerSPME fiber holder for CTC autosamplerVARISU57347ULog in för att kolla tillgänglighet och ert pris1 * 1 Items
Produktinformation
- Katalog #TillgänglighetPrice
- SPME stand, 15 ml
Produktinformation
- Katalog #TillgänglighetPrice
- SPME inlet guide for manual injectionSPME inlet guide for manual injectionVARISU57356ULog in för att kolla tillgänglighet och ert pris1 * 1 Items