Orders LinkContactUsLinkComponent
RapID™ System
RapID™ System
RapID™ Eric, 1
Katalog # OXOIR8323600
RapID™ System
Katalog # OXOIR8323600
CAS-nummer:  
RapID™ System
RapID™ System
RapID™ Eric, 1
CAS-nummer:  
Price
414,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
RapID™ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapID™ panels offer easy, one-step, simultaneous inoculation of dehydrated reactants in tray wells to identify anaerobes, coryneform Gram-positive bacilli, enterics, nonfermenters, streptococci, and yeast all within 4 hours. Systems are ideal for manual testing or as confirmation for automated test systems.
  • Rapid, accurate identification of over 400 medically important organisms
  • Simple, one-step inoculation
  • Four-hour aerobic incubation
  • Easy-to-read colour reactions
Documents
Specifikationer
Specifikationer
  • Förp.:
    1
  • Beskrivning:
    RapID™ Eric