Orders LinkContactUsLinkComponent
Blocking solution, BLOK™ Casein
Blocking solution, BLOK™ Casein
BLOK™ Casein in TBS, 1% solution, 2
Katalog # 786-196
Leverantör:   G-Biosciences
Blocking solution, BLOK™ Casein
Katalog # 786-196
Leverantör:  G-Biosciences
CAS-nummer:  
Blocking solution, BLOK™ Casein
Blocking solution, BLOK™ Casein
BLOK™ Casein in TBS, 1% solution, 2
CAS-nummer:  
Pris
2 155,00 kr
Log in för tillgänglighet
1 * 2 Items
Produktdetaljer och dokument
BLOK™ Casein is a purified protein, used for blocking free binding sites in Western blot, ELISA plate well, immunohistochemistry and other immunochemical applications.
  • Ready to use
  • Available in the choices of TBS or PBS buffers
Specifikationer
Specifikationer
  • Förp.:
    2
  • Beskrivning:
    BLOK™ Casein in TBS, 1% solution
Specifikationer