Xylen, J.T.Baker®
Plastic bottle, 5 L
Supplier: Avantor
Jump to:
Product Details & Documents
Synonyms: Dimethylbenzene
- CAS Number:1330-20-7
- Formula:C₆H₄(CH₃)₂
- MDL Number:MFCD00077264
- Molecular Weight:106.17 g/mol
- Merck Index:13,10136
- Boiling Pt:140 °C (1013 hPa)
- Melting Pt:–34 °C
- Density:0,86…0.88 g/cm³ (20 °C)
- Flash Pt:25 °C
- Storage Temperature:Omgivande
- EINECS:215-535-7
- UN:1307
- ADR:3,III
Documents
Specifications
Specification Test Results
- Benzene:max. 0.02 %
- Color (APHA):max. 20
- Density (g/ml) at 20°C:0.862 - 0.872
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



