Orders LinkContactUsLinkComponent
Boxes, with Hinged Lid
Boxes, with Hinged Lid
without label holder, 185×140×80 mm
Katalog # SEMA0341
Leverantör:   SEMADENI
Boxes, with Hinged Lid
Katalog # SEMA0341
Leverantör:  SEMADENI
Boxes, with Hinged Lid
Boxes, with Hinged Lid
without label holder, 185×140×80 mm
Price
255,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
The practical hinged flip-top box container is made from dimensionally stable Polypropylene. The standard colour is grey and the range includes a total of five different sizes. With lid and container made from a single block, snap-on cap and integral hinge, you can package your small item safely and securely.
  • Heavyweight boxes
  • Rigid and robust
  • Stackable
  • PP, grey
Specifikationer
  • Typ:
    without label holder
  • B×D×H:
    185×140×80 mm