Test tubes without rim, medium wall
50 mm, 12 mm
Katalog # MURRG002/75
Leverantör: MURRAY S
Gå till:
Produktdetaljer och dokument
Soda-lime glass, rimless.
- Good chemical and physical properties
- Medium-walled
Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.
Specifikationer
- Ø utv.:12 mm
- Längd:50 mm
- Seq:03