Filtertrattar
75 ml, 45 mm, 1
Katalog # LENZ07195134
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifikationer
- Platt Ø:45 mm
- Porositet:1
- Kapacitet:75 ml
- Seq:05