Orders LinkContactUsLinkComponent
Urglas
Urglas
Watch glass, PTFE, 126,36 mm
Katalog # KART646
Leverantör:   KARTELL
Urglas
Katalog # KART646
Leverantör:  KARTELL
Urglas
Urglas
Watch glass, PTFE, 126,36 mm
Price
630,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Made of PTFE, white.
  • Very good chemical, heat and mechanical resistance
  • Heat resistant up to +120 °C
  • For use with foodstuff
For blends or for covering vessels.
Specifikationer
  • Ø:
    126,36 mm
  • Beskrivning:
    Watch glass, PTFE
  • Seq:
    04