Density bottles, adjusted
100 ml
Katalog # 641-0900
Leverantör: Lenz Laborglas
Gå till:
Produktdetaljer och dokument
Borosilicate glass 3.3. DURAN®, Gay-Lussac type.
- The density bottles (pycnometers) are individually engraved with the actual capacity, with an accuracy of 0,001 cm³
- The top of stopper is ground and polished for ease of wiping clean
- Stopper is size NS 10/19 with capillary
* DIN ISO 3507
Dokument
BSE TSE Glass Columns
BSE/TSE-certifikat
Specifikationer
- Volym (nominell):100 ml