Dimroth-kylare
250 mm, 45/40 NS, 45/40 NS
Katalog # LENZ05240316
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For condensing vapours. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikat
Specifikationer
- Kärna:45/40 NS
- Mantellängd:250 mm
- NS-hylsa:45/40 NS
- Seq:07



