Dimroth-kylare
250 mm, 24/29 NS, 24/29 NS
Katalog # LENZ05240313
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For condensing vapours. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikat
Specifikationer
- Kärna:24/29 NS
- Mantellängd:250 mm
- NS-hylsa:24/29 NS
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



