Orders LinkContactUsLinkComponent
Sleeve Sampler
Sleeve Sampler
Heavy duty handle, Rostfritt stål 316L, 1000 mm
Katalog # SAMP1035A-1000
Leverantör:   Sampling Systems
Sleeve Sampler
Katalog # SAMP1035A-1000
Leverantör:  Sampling Systems
Sleeve Sampler
Sleeve Sampler
Heavy duty handle, Rostfritt stål 316L, 1000 mm
Price
3 600,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
The sleeve sampler is specially designed for large volumes at great depths.
  • Adjustable length and bright finish design
  • Extremely strong and robust extension rods
Suitable for free-flowing powders and granules.
Specifikationer
  • Beskrivning:
    Heavy duty handle
  • Längd:
    1000 mm
  • Material:
    Rostfritt stål 316L
  • Seq:
    04