N,N-Dimetylformamid ≥99%
Glass bottle, 10.000 mL
Leverantör: Thermo Fisher Scientific
N,N-Dimetylformamid ≥99%
Katalog # A13547.0D
Leverantör: Thermo Fisher Scientific
CAS-nummer: 68-12-2
Gå till:
Product Details & Documents
Purity: 99% CAS No.: 68-12-2 Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 Flash Point: 57°(135°F) Refractive Index: 1.4310 Odour: Amine-like pH: 7 Melting Point: -61° Boiling Point: 153° Beilstein Reference: 605365 Merck Reference: 14,3243 UN No.: 2265 MDL No.: MFCD00003284
Synonymer: DMF
- CAS-nummer:68-12-2
- Formel:HCON(CH₃)₂
- MDL-nummer:MFCD00003284
- Molecular Weight:73.09 g/mol
- Merck Index:14,3243
- Kokande Pt:153 °C
- Smält Pt:–61 °C
- Densitet:0,944
- Flash Pt:57 °C (135 °F)
- Förvaringstemperatur:Omgivande
- EINECS:200-679-5
- FN:2265
- ADR:3,III
- REACH:01-2119475605-32
Documents
Specifikationer
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:10.000 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



