Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Ammoniumformat, (max. 3% H₂O) 97%

Glass bottle for solids, 250 g
Katalog # A10699.30
Irriterande
Leverantör:   Thermo Fisher Scientific
Ammoniumformat, (max. 3% H₂O) 97%
Katalog # A10699.30
CAS-nummer:  540-69-2
Ammoniumformat, (max. 3% H₂O) 97%
Ammoniumformat, (max. 3% H₂O) 97%

Glass bottle for solids, 250 g

CAS-nummer:  540-69-2
Price
490,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 250 g
Product Details & Documents

Purity: 97% CAS No.: 540-69-2 Molecular Formula: CH5NO2 Formula Weight: 63.06 Odour: Ammonia-like Melting Point: ca 116° Beilstein Reference: 3625095 Merck Reference: 14,523 MDL No.: MFCD00013103

Synonymer:  Formic acid ammonium salt
  • CAS-nummer:540-69-2
  • Formel:HCO₂NH₄
  • MDL-nummer:MFCD00013103
  • Molecular Weight:63.06 g/mol
  • Merck Index:14,523
  • Smält Pt:∼116 °C
  • Densitet:1,28
  • Densitet:1.28
  • Förvaringstemperatur:Omgivande
  • EINECS:208-753-9
Specifikationer

Specifikationer

  • Förpackningstyp:
    Glass bottle for solids
  • Förp.:
    250 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.