Bottles, narrow or wide neck, square or rectangular, with thread
2000 ml, 80 mm, 90,5×125,0×230,0 mm
Catalog # RIXI2-M0-2000-80
Supplier: BERLIN PACKAGING RIXIUS GMBH
Bottles, narrow or wide neck, square or rectangular, with thread
Catalog # RIXI2-M0-2000-80
Supplier: BERLIN PACKAGING RIXIUS GMBH
Jump to:
Product Details & Documents
HDPE, natural.
- Rigid and shatter-proof
- Resistant to water, aqueous solutions and most chemicals
- Excellent water vapour resistance
- Ribbed sides for secure grip
Square or rectangular bottles with large filling opening for packaging and storage of liquids, powders and granulates.
Ordering information: Screw caps must be ordered seperately.
Documents
RIXI2-E0-1000-40_TDS_EN
Datasheet
Specifications
- Kapacitet:2000 ml
- Seq:20
- B×D×H:90,5×125,0×230,0 mm
- Hals, innerdiam.:80 mm