Orders LinkContactUsLinkComponent
Caps with dropper inserts for bottles
Caps with dropper inserts for bottles
Bottles with GL 32 thread
Katalog # VITL83308
Leverantör:   VITLAB
Caps with dropper inserts for bottles
Katalog # VITL83308
Leverantör:  VITLAB
Caps with dropper inserts for bottles
Caps with dropper inserts for bottles
Bottles with GL 32 thread
Price
780,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 12 Items
Produktdetaljer och dokument
Made from HDPE.
  • Extra long, fine dropping tip for accurate dispensing
  • Good chemical resistance
  • Good temperature resistance
For bottles with GL threads. Cap with dropper insert, complete with screw cap.
Specifikationer
  • För:
    Bottles with GL 32 thread
  • Seq:
    03