Washers, BOLA
for tubing 1,6 mm I-Ø/3,2 mm O-Ø, PA
Katalog # BOHLF728-16
Leverantör: Bohlender
Gå till:
Produktdetaljer och dokument
PA
- Temperature resistance: from –40 to +100 °C
For stabilising the flange and avoiding damages when tightening the tube end fitting.
Specifikationer
- Beskrivning:for tubing 1,6 mm I-Ø/3,2 mm O-Ø
- Material:PA