O-Ring seals for flat flanges
planflänsar DN 60
Catalog # LENZ06248206
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Silicone or Silicone PTFE-coated.
- Excellent chemical resistance
- Excellent temperature resistance
Seals for flat flanges LF 60-200.
Specifications
- För:planflänsar DN 60
- Seq:05