Sample cuvettes
Sample cuvettes, 1,5 ml, 10 mm, PS
Katalog # HACH2629500
Leverantör: Hach
Gå till:
Product Details & Documents
Sample cuvettes for photometric measurements.
- Available with or without lid
- Available reusable (made from glass) or for single use (made from plastics)
Specifikationer
- Skiktdjup:10 mm
- Volym:1,5 ml
- Material:PS
- Beskrivning:Sample cuvettes
- Seq:01