Skruvlock, Nalgene®
Linerless, 28-415, Brun, PP
215-1621
PP, färgat.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
- Lock storlek:28-415
- Färg:Brun
- Beskrivning:Linerless
- Material:PP
- Seq:29
- Cat. No.:215-1621
- Miljöcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact