Skruvlock, Nalgene®
Linerless, 28-415, Röd, PP
Katalog # 215-1608
Leverantör: Thermo Fisher Scientific
Skruvlock, Nalgene®
Katalog # 215-1608
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
PP, färgat.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifikationer
- Lock storlek:28-415
- Färg:Röd
- Beskrivning:Linerless
- Material:PP
- Seq:19
- Cat. No.:215-1608
- Miljöcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact