Skruvlock, Nalgene®
Linerless, 24-415, Ofärgad, PP
215-1598
PP, färgat.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
- Lock storlek:24-415
- Färg:Ofärgad
- Beskrivning:Linerless
- Material:PP
- Seq:03
- Cat. No.:215-1598
- Miljöcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact