Insulin, Reference Standards
Produktdetaljer och dokument
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Insulin Glargine for Peak Identication: Mixture of Insulin Glargine and 0A-Arg-Insulin Glargine
Certifications: US Pharmacopea Reference Standard
Varning: Cold Shipment required
Dokument
Certificates
Sök efter LOT-nummerSpecifikationer
Specifikationer
- Förpackningstyp:Vial
- Förp.:50 mg
- Standard:USP reference standard
- Beskrivning:Insulin Pork
- Cat. No.:1342300.
- CAS-nr:12584-58-6
- Benämning:Insulin Pork