Orders LinkContactUsLinkComponent
Gadodiamide, Reference Standards
Gadodiamide, Reference Standards
Gadodiamide Related Compound B, USP reference standard, Vial, 50 mg
Katalog # 1287529.
Leverantör:   USP
Gadodiamide, Reference Standards
Katalog # 1287529.
Leverantör:  USP
CAS-nummer:  
Gadodiamide, Reference Standards
Gadodiamide Related Compound B, USP reference standard, Vial, 50 mg
CAS-nummer:  
Pris
13 400,00 kr
Log in för tillgänglighet
1 * 50 mg
Produktdetaljer och dokument
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifikationer
Specifikationer
  • Förpackningstyp:
    Vial
  • Förp.:
    50 mg
  • Standard:
    USP reference standard
  • Beskrivning:
    Gadodiamide Related Compound B
  • Cat. No.:
    1287529.
  • Benämning:
    Gadodiamide Related Compound B
  • Synonym(s):
    gadolinium disodium diethylenetriamine pentaacetic acid
Specifikationer