Weighing scoops
3 ml, 82 mm
Katalog # 611-9309
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
Borosilicate glass.
- Ideal for weighing small samples
- The tubular stem of the scoop also works as a weighing funnel and facilitates emptying of the scoop
Documents
BSE TSE Glass Columns
BSE/TSE-certifikat
Specifikationer
- Kapacitet:3 ml
- Längd:82 mm