ACES
Glass bottle for solids, 1 kg
Katalog # MOLE14774016-1KG
Marketsource
Leverantör: Molekula
Gå till:
Product Details & Documents
Synonymer: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
- CAS-nummer:7365-82-4
- Formel:H₂NCOCH₂NHCH₂CH₂SO₃H
- MDL-nummer:MFCD00008030
- Molecular Weight:182.2 g/mol
- Merck Index:13,00037
- Smält Pt:270…295 °C
- Förvaringstemperatur:Omgivande
- EINECS:230-908-4
Specifikationer
Specifikationer
- Förpackningstyp:Glass bottle for solids
- Förp.:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



