O-Ring seals for flat flanges
planflänsar DN 100
Katalog # LENZ06248210
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
Silicone or Silicone PTFE-coated.
- Excellent chemical resistance
- Excellent temperature resistance
Seals for flat flanges LF 60-200.
Specifikationer
- För:planflänsar DN 100
- Seq:06