Urinary system model
Dual Sex Urinary System, 6 part
Katalog # BINH1000317
Marketsource
Leverantör: 3B Scientific
Gå till:
Produktdetaljer och dokument
Urinary system all-in-one model on baseboard showing:
- Structures of retroperitoneal cavity
- Large and small pelvis with bones and muscles
- Inferior vena cava
- Aorta with its branches including iliacal vessels
- Upper urinary tract
- Rectum
- Kidney with adrenal gland
The prostate model provides a cross section of the male genital organs showing a healthy prostate with bladder, urethra, testicle, symphysis and rectum.
Specifikationer
- Beskrivning:Dual Sex Urinary System, 6 part
Rekommendationer
Rekommendationer anpassas endast baserat på dina shoppingpreferenser om du har gett ditt samtycke genom att aktivera "Förbättra min shoppingupplevelse" på sidan "Personlig information".
Annars får du generiska rekommendationer.