Calibration weights, cylindrical, OIML, class E2
500 g, 0,8 ± mg, Cylindrical test weight
Catalog # SARBYCW5528-00
Supplier: Sartorius Balances
Calibration weights, cylindrical, OIML, class E2
Catalog # SARBYCW5528-00
Supplier: Sartorius Balances
Jump to:
Product Details & Documents
Class E2 cyclindrical calibration weights.
Specifications
- Vikt:500 g
- Seq:10
- Tolerans:0,8 ± mg
- Typ:Cylindrical test weight