Calibration weights, cylindrical, OIML, class E2
200 g, 0,30 ± mg, Cylindrical test weight
Katalog # SARBYCW5228-00
Leverantör: Sartorius Balances
Calibration weights, cylindrical, OIML, class E2
Katalog # SARBYCW5228-00
Leverantör: Sartorius Balances
Gå till:
Product Details & Documents
Class E2 cyclindrical calibration weights.
Specifikationer
- Vikt:200 g
- Seq:09
- Tolerans:0,30 ± mg
- Typ:Cylindrical test weight