Calibration weights, cylindrical, OIML, class E2
10 g, 0,060 ± mg, Cylindrical test weight
Katalog # SARBYCW4128-00
Leverantör: Sartorius Balances
Calibration weights, cylindrical, OIML, class E2
Katalog # SARBYCW4128-00
Leverantör: Sartorius Balances
Gå till:
Produktdetaljer och dokument
Class E2 cyclindrical calibration weights.
Specifikationer
- Vikt:10 g
- Seq:04
- Tolerans:0,060 ± mg
- Typ:Cylindrical test weight