Calibration weights, cylindrical, OIML, class E2
50 g, 0,10 ± mg, Cylindrical test weight
Katalog # SARBYCW4528-00
Leverantör: Sartorius Balances
Calibration weights, cylindrical, OIML, class E2
Katalog # SARBYCW4528-00
Leverantör: Sartorius Balances
Gå till:
Product Details & Documents
Class E2 cyclindrical calibration weights.
Specifikationer
- Vikt:50 g
- Seq:06
- Tolerans:0,10 ± mg
- Typ:Cylindrical test weight