Vacuum stopcocks
Vacuum stopcocks, two way, right-angled, 8,0 mm, 24 NS
Katalog # LENZ02962408
Leverantör: Lenz Laborglas
Gå till:
Produktdetaljer och dokument
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
- Excellent chemical resistance
- Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Specifikationer
- Hål Ø.:8,0 mm
- Beskrivning:Vacuum stopcocks, two way, right-angled
- Seq:19
- Kran:24 NS