VWR®, Spreaders, Triangle Shaped, Disposable
Steril, Blå, Individuellt
Leverantör: VWR Collection
Gå till:
Product Details & Documents
PP, sterile (individually wrapped) or non sterile (bulk packed). Convenient and economical, these cell spreaders are designed to simplify bacterial culture work. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
- Durable enough to be autoclaved and used again
- Width: 30 mm (blue)/60 mm (yellow)
Documents
Certificates
Sök efter LOT-nummerCoC VWR, Spreaders, Triangle Shaped, Disposable.pdf
Försäkran om överensstämmelse
Specifikationer
- Förp.:Individuellt
- Färg:Blå
- Beskrivning:Steril
- Seq:01