Orders LinkContactUsLinkComponent
VWR®, Spreaders, Triangle Shaped, Disposable
VWR®, Spreaders, Triangle Shaped, Disposable
Osteril, Blå, Bulk
Katalog # 612-2687
Leverantör:   VWR Collection
VWR®, Spreaders, Triangle Shaped, Disposable
Katalog # 612-2687
Leverantör:  VWR Collection
VWR®, Spreaders, Triangle Shaped, Disposable
VWR®, Spreaders, Triangle Shaped, Disposable
Osteril, Blå, Bulk
Price
740,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 25 Items
Product Details & Documents
PP, sterile (individually wrapped) or non sterile (bulk packed). Convenient and economical, these cell spreaders are designed to simplify bacterial culture work. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
  • Durable enough to be autoclaved and used again
  • Width: 30 mm (blue)/60 mm (yellow)
Specifikationer
  • Förp.:
    Bulk
  • Färg:
    Blå
  • Beskrivning:
    Osteril
  • Seq:
    03