Orders LinkContactUsLinkComponent
ChromBox kit
ChromBox kit
Tubing clip
Catalog # JOURJR-9001
Supplier:   VICI JOUR RESEARCH
ChromBox kit
Catalog # JOURJR-9001
Supplier:  VICI JOUR RESEARCH
ChromBox kit
Tubing clip
Price
66,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.
Ordering information: Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.
Specifications
  • Beskrivning:
    Tubing clip
  • Seq:
    07