ChromBox kit
ChromBox kit
Katalog # JOURJR-34N
Leverantör: VICI JOUR RESEARCH
Gå till:
Produktdetaljer och dokument
The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.
: Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.
Specifikationer
- Beskrivning:ChromBox kit
- Seq:01