Order Entry
Sweden
Orders LinkContactUsLinkComponent
ChromBox kit

ChromBox kit

Tubing elbow, 90°
Katalog # JOURJR-357090
Leverantör:   VICI JOUR RESEARCH
ChromBox kit
Katalog # JOURJR-357090
Leverantör:  VICI JOUR RESEARCH
ChromBox kit

Tubing elbow, 90°

Price
46,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument

The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.

Ordering information: Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.

Specifikationer
  • Beskrivning:
    Tubing elbow, 90°
  • Seq:
    02