ChromBox kit
Tubing elbow, 180°
Katalog # JOURJR-357180
Leverantör: VICI JOUR RESEARCH
Gå till:
Produktdetaljer och dokument
The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.
Ordering information: Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.
Specifikationer
- Beskrivning:Tubing elbow, 180°
- Seq:03