Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Slimsette®, M510
Biopsy processing/embedding cassettes, Slimsette®, M510
Blå
Katalog # 720-1609
Leverantör:   Simport Scientific
Biopsy processing/embedding cassettes, Slimsette®, M510
Katalog # 720-1609
Leverantör:  Simport Scientific
Biopsy processing/embedding cassettes, Slimsette®, M510
Biopsy processing/embedding cassettes, Slimsette®, M510
Blå
Price
1 675,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 500 Items
Produktdetaljer och dokument
Acetal polymer. Cassette design is similar to that of the M509 model but specially designed to hold biopsy specimens.
  • Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
  • Disposable plastic cassettes are highly resistant to histological solutions
  • Small, square openings for good fluid exchange
  • Integral, recessed lid
  • Front face inclined 45° to enable use with certain automatic labelling systems
W×D×H: 41×28,5×6 mm
Packaging: Each case contains three boxes holding 500 cassettes each.
Specifikationer
  • Färg:
    Blå